Assay Details
Target Gene Details
Entrez Gene ID: | 116362 |
Gene Name: | retinol binding protein 7 |
Gene Aliases: |
CRBP4, CRBPIV |
Location: |
Chr.1:9997197-10016020 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBP7 | NM_052960.2 | NP_443192.1 | ||
| AF399927.1 | AAK85409.1 | |||
| AK312027.1 | ||||
| AY145438.1 | AAN61071.1 | |||
| BC033883.1 | AAH33883.1 | |||
| BC063013.1 | AAH63013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2664618 | Chr.1:10007923 - 10012254 on Build GRCh38 | Deletion |
|
| esv2671412 | Chr.1:10007064 - 10008732 on Build GRCh38 | Deletion |
|
| esv3585193 | Chr.1:10007132 - 10008617 on Build GRCh38 | Loss |
|
| nsv1001742 | Chr.1:9911718 - 10056709 on Build GRCh38 | Gain |
|
| esv3585194 | Chr.1:10007990 - 10012138 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map