Assay Details
Target Gene Details
Entrez Gene ID: | 81621 |
Gene Name: | Kazal type serine peptidase inhibitor domain 1 |
Gene Aliases: |
BONO1, FKSG28, FKSG40, IGFBP-rP10 |
Location: |
Chr.10:101061141-101068141 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAZALD1 | NM_001319303.1 | NP_001306232.1 | ||
| NM_030929.4 | 5 | 2238 | NP_112191.2 | |
| NR_135067.1 | ||||
| XM_017016714.1 | XP_016872203.1 | |||
| XM_017016715.1 | XP_016872204.1 | |||
| AF333487.1 | AAG50291.1 | |||
| AK172862.1 | 5 | 1489 | BAD18818.1 | |
| AK172864.1 | ||||
| AK225949.1 | ||||
| AY014271.1 | AAG50354.1 | |||
| AY359087.1 | 5 | 1494 | AAQ89445.1 | |
| BC007758.2 | 5 | 1639 | AAH07758.1 | |
| BG034107.1 | ||||
| CX871488.1 | ||||
| DA422973.1 | 1 | 123 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831965 | Chr.10:100916724 - 101082796 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map