Assay Details
Target Gene Details
Entrez Gene ID: | 55731 |
Gene Name: | family with sequence similarity 222 member B |
Gene Aliases: |
C17orf63 |
Location: |
Chr.17:28755978-28855036 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM222B | NM_001077498.2 | 3 | 824 | NP_001070966.1 |
| NM_001288631.1 | 4 | 995 | NP_001275560.1 | |
| NM_001288632.1 | 5 | 906 | NP_001275561.1 | |
| NM_001288633.1 | 4 | 900 | NP_001275562.1 | |
| NM_001288634.1 | 3 | 893 | NP_001275563.1 | |
| NM_001288635.1 | 4 | 872 | NP_001275564.1 | |
| NM_001288636.1 | 6 | 896 | NP_001275565.1 | |
| NM_001288637.1 | 5 | 862 | NP_001275566.1 | |
| NM_001288638.1 | 5 | 794 | NP_001275567.1 | |
| NM_001288639.1 | 4 | 746 | NP_001275568.1 | |
| NM_001288640.1 | 3 | 712 | NP_001275569.1 | |
| NM_018182.3 | 4 | 858 | NP_060652.2 | |
| XM_011524986.2 | 3 | 789 | XP_011523288.1 | |
| XM_011524987.1 | 4 | 624 | XP_011523289.1 | |
| XM_011524988.2 | 3 | 840 | XP_011523290.1 | |
| XM_017024831.1 | 4 | 948 | XP_016880320.1 | |
| XM_017024832.1 | 3 | 1213 | XP_016880321.1 | |
| XM_017024833.1 | 3 | 753 | XP_016880322.1 | |
| XM_017024834.1 | 3 | 951 | XP_016880323.1 | |
| XM_017024835.1 | 4 | 1150 | XP_016880324.1 | |
| AK000147.1 | BAA90974.1 | |||
| AK001562.1 | 4 | 809 | BAA91757.1 | |
| AK057665.1 | 3 | 808 | ||
| AK095569.1 | 3 | 893 | ||
| AK299477.1 | 3 | 658 | ||
| BC021228.2 | 3 | 708 | AAH21228.1 | |
| BX647766.1 | 3 | 643 |

Genomic Map