Assay Details
Target Gene Details
Entrez Gene ID: | 64429 |
Gene Name: | zinc finger DHHC-type containing 6 |
Gene Aliases: |
DHHC-6, ZNF376 |
Location: |
Chr.10:112424411-112448335 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZDHHC6 | NM_001303134.1 | 2 | 633 | NP_001290063.1 |
| NM_022494.2 | 2 | 633 | NP_071939.1 | |
| XM_005270051.4 | 2 | 1568 | XP_005270108.1 | |
| XM_006717948.3 | 2 | 739 | XP_006718011.1 | |
| XM_006717949.3 | 2 | 630 | XP_006718012.1 | |
| XM_011540091.2 | 2 | 621 | XP_011538393.1 | |
| XM_011540092.2 | 2 | 630 | XP_011538394.1 | |
| XM_011540093.1 | 2 | 739 | XP_011538395.1 | |
| XM_011540094.2 | 2 | 610 | XP_011538396.1 | |
| XM_011540095.2 | 2 | 610 | XP_011538397.1 | |
| XM_017016561.1 | 2 | 559 | XP_016872050.1 | |
| XM_017016562.1 | 2 | 739 | XP_016872051.1 | |
| XM_017016563.1 | 2 | 552 | XP_016872052.1 | |
| XM_017016564.1 | 2 | 739 | XP_016872053.1 | |
| XM_017016565.1 | 2 | 805 | XP_016872054.1 | |
| XM_017016566.1 | 2 | 630 | XP_016872055.1 | |
| XM_017016567.1 | 2 | 630 | XP_016872056.1 | |
| XM_017016568.1 | 2 | 536 | XP_016872057.1 | |
| AF267740.1 | 2 | 588 | AAL56663.1 | |
| AK025605.1 | 2 | 588 | BAB15187.1 | |
| AK223086.1 | 2 | 587 | BAD96806.1 | |
| AK308026.1 | 2 | 781 | ||
| BC007213.1 | 2 | 575 | AAH07213.1 | |
| BC017434.2 | 2 | 575 | AAH17434.1 | |
| BX647270.1 | 2 | 498 | ||
| DA745043.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831990 | Chr.10:112349267 - 112531997 on Build GRCh38 | Loss |
|
| nsv831991 | Chr.10:112413098 - 112582139 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map