Assay Details
Target Gene Details
Entrez Gene ID: | 26268 |
Gene Name: | F-box protein 9 |
Gene Aliases: |
FBX9, NY-REN-57, VCIA1, dJ341E18.2 |
Location: |
Chr.6:53057807-53100873 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO9 | NM_012347.4 | NP_036479.1 | ||
| NM_033480.2 | NP_258441.1 | |||
| NM_033481.3 | NP_258442.2 | |||
| XM_005248995.4 | XP_005249052.1 | |||
| XM_005248996.4 | XP_005249053.1 | |||
| AB209833.1 | BAD93070.1 | |||
| AF174597.1 | AAF04518.1 | |||
| AF176704.1 | AAF03704.1 | |||
| AK021559.1 | ||||
| AL137520.1 | CAB70786.2 | |||
| AY858798.1 | AAW51115.1 | |||
| BC000650.2 | AAH00650.2 |
More Information
Set Membership: |
|

Genomic Map