Assay Details
Target Gene Details
Entrez Gene ID: | 11000 |
Gene Name: | solute carrier family 27 member 3 |
Gene Aliases: |
ACSVL3, FATP3, VLCS-3 |
Location: |
Chr.1:153774989-153780157 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC27A3 | NM_001317929.1 | 1 | 124 | NP_001304858.1 |
| NM_024330.1 | 1 | 124 | NP_077306.1 | |
| AJ577572.1 | 1 | 417 | CAE12158.1 | |
| AK027499.1 | BAB55156.1 | |||
| AK074134.1 | 1 | 97 | BAB84960.1 | |
| AK075377.1 | 1 | 117 | BAC11578.1 | |
| AK123997.1 | 1 | 106 | ||
| AK125102.1 | 1 | 1062 | BAC86050.1 | |
| AY358409.1 | 1 | 112 | AAQ88775.1 | |
| BC003041.1 | 1 | 124 | AAH03041.1 | |
| BC009916.2 | 1 | 124 | AAH09916.1 | |
| BC029792.1 | AAH29792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv547941 | Chr.1:153736430 - 153828622 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map