Assay Details
Target Gene Details
Entrez Gene ID: | 84961 |
Gene Name: | F-box and leucine rich repeat protein 20 |
Gene Aliases: |
Fbl2, Fbl20 |
Location: |
Chr.17:39252644-39402659 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXL20 | NM_001184906.1 | 14 | 2135 | NP_001171835.1 |
| NM_032875.2 | 15 | 2231 | NP_116264.2 | |
| XM_005257746.3 | 15 | 2422 | XP_005257803.2 | |
| XM_005257747.3 | 14 | 2203 | XP_005257804.2 | |
| AK123048.1 | 15 | 2232 | ||
| AK291844.1 | ||||
| BC007557.1 | 15 | 2198 | AAH07557.2 | |
| DQ223959.1 | 14 | 2102 | ABB03906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055612 | Chr.17:39253612 - 39406983 on Build GRCh38 | Gain |
|
| nsv833441 | Chr.17:39164554 - 39320543 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map