Assay Details
Target Gene Details
Entrez Gene ID: | 153642 |
Gene Name: | arylsulfatase family member K |
Gene Aliases: |
TSULF |
Location: |
Chr.5:95555108-95605102 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARSK | NM_198150.2 | 1 | 158 | NP_937793.1 |
| XM_005271904.3 | 1 | 171 | XP_005271961.1 | |
| AK303855.1 | 1 | 210 | ||
| AK316253.1 | 1 | 178 | ||
| AY358596.1 | 1 | 118 | AAQ88959.1 | |
| AY875939.1 | AAW66667.1 | |||
| BC036047.1 | AAH36047.1 | |||
| BC050392.1 | 1 | 117 | AAH50392.1 | |
| BC130329.1 | 1 | 96 |
More Information
Set Membership: |
|

Genomic Map