Assay Details
Target Gene Details
Entrez Gene ID: | 5690 |
Gene Name: | proteasome subunit beta 2 |
Gene Aliases: |
HC7-I |
Location: |
Chr.1:35599541-35641844 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMB2 | NM_001199779.1 | 4 | 454 | NP_001186708.1 |
| NM_001199780.1 | 3 | 298 | NP_001186709.1 | |
| NM_002794.4 | 4 | 822 | NP_002785.1 | |
| AB209470.1 | 4 | 508 | BAD92707.1 | |
| AI025755.1 | 3 | 331 | ||
| AK296956.1 | 4 | 429 | ||
| AK310023.1 | 3 | 298 | ||
| AV762678.2 | 4 | 516 | ||
| BC101836.1 | 4 | 454 | AAI01837.1 | |
| BC105126.1 | 4 | 454 | AAI05127.1 | |
| BC107901.1 | 4 | 469 | AAI07902.1 | |
| BM849239.1 | 4 | 454 | ||
| BT007137.1 | 4 | 410 | AAP35801.1 | |
| CR456862.1 | 4 | 410 | CAG33143.1 | |
| D26599.1 | 4 | 439 | BAA05646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012556 | Chr.1:35413704 - 35703660 on Build GRCh38 | Gain |
|
| nsv433 | Chr.1:35582457 - 35627412 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map