Assay Details
Target Gene Details
Entrez Gene ID: | 9553 |
Gene Name: | mitochondrial ribosomal protein L33 |
Gene Aliases: |
C2orf1, L33mt, MRP-L33, RPL33L |
Location: |
Chr.2:27771717-27779741 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL33 | NM_004891.3 | 4 | 269 | NP_004882.1 |
| NM_145330.2 | 3 | 162 | NP_663303.1 | |
| AA836397.1 | 1 | 260 | ||
| AF047440.1 | 4 | 243 | AAC39891.1 | |
| BC009475.1 | 4 | 221 | AAH09475.1 | |
| BC020834.1 | 4 | 256 | AAH20834.1 | |
| BC090047.1 | 3 | 162 | AAH90047.1 | |
| BI601959.1 | 4 | 272 | ||
| CR456888.1 | CAG33169.1 | |||
| CR542117.1 | CAG46914.1 |
More Information
Set Membership: |
|

Genomic Map