Assay Details
Target Gene Details
Entrez Gene ID: | 146562 |
Gene Name: | chromosome 16 open reading frame 71 |
Gene Aliases: |
- |
Location: |
Chr.16:4734288-4749396 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C16orf71 | NM_139170.2 | 4 | 1099 | NP_631909.2 |
| XM_005255144.3 | 3 | 828 | XP_005255201.1 | |
| XM_017022975.1 | 4 | 955 | XP_016878464.1 | |
| XM_017022976.1 | 4 | 717 | XP_016878465.1 | |
| AF447587.1 | 4 | 1073 | AAM22870.1 | |
| AK125251.1 | 1 | 128 | ||
| AK301794.1 | 4 | 925 | ||
| BC035024.1 | 4 | 1049 | AAH35024.1 | |
| BX648126.1 | 4 | 885 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| nsv571302 | Chr.16:4708464 - 4762815 on Build GRCh38 | Gain |
|
| nsv1050237 | Chr.16:4641028 - 4744872 on Build GRCh38 | Loss |
|

Genomic Map