Assay Details
Target Gene Details
Entrez Gene ID: | 475 |
Gene Name: | antioxidant 1 copper chaperone |
Gene Aliases: |
ATX1, HAH1 |
Location: |
Chr.5:151742822-151758649 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATOX1 | NM_004045.3 | 3 | 236 | NP_004036.1 |
| AK128210.1 | 3 | 1113 | ||
| AK293063.1 | 3 | 243 | ||
| BC112248.1 | 3 | 318 | AAI12249.1 | |
| BC112250.1 | 3 | 318 | AAI12251.1 | |
| BI599233.1 | 3 | 244 | ||
| BT009786.1 | 2 | 138 | AAP88788.1 | |
| BU172675.1 | 3 | 614 | ||
| CN280769.1 | 3 | 213 | ||
| CR456901.1 | 2 | 138 | CAG33182.1 |
More Information
Set Membership: |
|

Genomic Map