Assay Details
Target Gene Details
Entrez Gene ID: | 9175 |
Gene Name: | mitogen-activated protein kinase kinase kinase 13 |
Gene Aliases: |
LZK, MEKK13, MLK |
Location: |
Chr.3:185282938-185489094 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K13 | NM_001242314.1 | 15 | 3354 | NP_001229243.1 |
| NM_001242317.1 | 13 | 2618 | NP_001229246.1 | |
| NM_004721.4 | 14 | 3469 | NP_004712.1 | |
| XM_011513310.2 | 14 | 3627 | XP_011511612.1 | |
| XM_017007456.1 | 15 | 3365 | XP_016862945.1 | |
| XM_017007457.1 | 13 | 3067 | XP_016862946.1 | |
| XM_017007458.1 | 15 | 3255 | XP_016862947.1 | |
| XM_017007459.1 | 14 | 3194 | XP_016862948.1 | |
| AB001872.1 | 14 | 3335 | BAA24817.1 | |
| AK297646.1 | 13 | 2606 | ||
| AK302951.1 | 14 | 2831 | ||
| BC111726.1 | AAI11727.1 |
Target Gene Details
Entrez Gene ID: | 90407 |
Gene Name: | transmembrane protein 41A |
Gene Aliases: |
2900010K02Rik |
Location: |
Chr.3:185482671-185499057 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM41A |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829818 | Chr.3:185411745 - 185567640 on Build GRCh38 | Loss |
|
| esv3893837 | Chr.3:185291669 - 185483550 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map