Assay Details
Target Gene Details
Entrez Gene ID: | 7164 |
Gene Name: | tumor protein D52-like 1 |
Gene Aliases: |
D53 |
Location: |
Chr.6:125153483-125264407 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPD52L1 | NM_001003396.2 | 1 | 147 | NP_001003396.1 |
| NM_001003397.2 | 1 | 147 | NP_001003397.1 | |
| NM_001300994.2 | 1 | 147 | NP_001287923.1 | |
| NM_001318903.1 | 1 | 147 | NP_001305832.1 | |
| NM_003287.3 | 1 | 147 | NP_003278.1 | |
| XM_006715557.3 | 1 | 393 | XP_006715620.1 | |
| XM_017011237.1 | 1 | 393 | XP_016866726.1 | |
| XM_017011238.1 | 1 | 393 | XP_016866727.1 | |
| AF208012.1 | AAG49634.1 | |||
| AK291866.1 | 1 | 111 | ||
| AK291872.1 | ||||
| BC002375.2 | AAH02375.1 | |||
| BE409865.1 | 1 | 174 | ||
| BM546629.1 | ||||
| BQ641312.1 | ||||
| CB114736.1 | 1 | 147 | ||
| U44427.1 | 1 | 104 | AAB40894.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830795 | Chr.6:125056686 - 125260117 on Build GRCh38 | Loss |
|
| nsv1029255 | Chr.6:124927961 - 125234814 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map