Assay Details
Target Gene Details
Entrez Gene ID: | 332 |
Gene Name: | baculoviral IAP repeat containing 5 |
Gene Aliases: |
API4, EPR-1 |
Location: |
Chr.17:78214196-78225635 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BIRC5 | NM_001012270.1 | NP_001012270.1 | ||
| NM_001012271.1 | NP_001012271.1 | |||
| NM_001168.2 | NP_001159.2 | |||
| AB028869.1 | BAA93676.1 | |||
| AB154416.1 | BAD11155.1 | |||
| AF077350.1 | AAD34226.1 | |||
| AK223428.1 | BAD97148.1 | |||
| AK301793.1 | ||||
| AK311917.1 | ||||
| AL547142.3 | ||||
| AY927772.1 | 2 | 237 | AAY15202.1 | |
| BC008718.2 | AAH08718.1 | |||
| BC034148.1 | AAH34148.1 | |||
| BC065497.1 | AAH65497.1 | |||
| BE537971.1 | ||||
| BG178974.1 | ||||
| BI520186.1 | ||||
| BQ687404.1 | ||||
| CB112371.1 | ||||
| CR541740.1 | CAG46540.1 | |||
| DQ227257.1 | ||||
| DQ310375.1 | ABC42341.1 | |||
| DQ310376.1 | ABC42342.1 | |||
| DQ310377.1 | ABC42343.1 | |||
| DQ310378.1 | ABC42344.1 | |||
| DQ310379.1 | ABC42345.1 | |||
| DQ508245.1 | ||||
| DQ508246.1 | ||||
| DQ508247.1 | ||||
| DQ508248.1 | ||||
| DQ508249.1 | ||||
| HM625836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524306 | Chr.17:78183449 - 78280693 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map