Assay Details
Target Gene Details
Entrez Gene ID: | 84902 |
Gene Name: | centrosomal protein 89 |
Gene Aliases: |
CCDC123, CEP123 |
Location: |
Chr.19:32875925-32972029 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP89 | NM_032816.4 | 14 | 1596 | NP_116205.3 |
| XM_005259344.3 | 14 | 1519 | XP_005259401.1 | |
| XM_011527425.2 | 11 | 1043 | XP_011525727.1 | |
| XM_017027398.1 | 14 | 1591 | XP_016882887.1 | |
| AK027546.1 | 13 | 1297 | BAB55190.1 | |
| AL832158.1 | 13 | 1419 | ||
| BC136328.1 | 14 | 1511 | ||
| BU196203.1 | 6 | 573 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893199 | Chr.19:32901584 - 32957298 on Build GRCh38 | Gain |
|
| nsv833807 | Chr.19:32889838 - 33077690 on Build GRCh38 | Loss |
|
| nsv1060014 | Chr.19:32854920 - 33011720 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map