Assay Details
Target Gene Details
Entrez Gene ID: | 5876 |
Gene Name: | Rab geranylgeranyltransferase beta subunit |
Gene Aliases: |
GGTB |
Location: |
Chr.1:75786194-75795090 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RABGGTB | NM_004582.3 | NP_004573.2 | ||
| NR_073562.1 | ||||
| AB209023.1 | BAD92260.1 | |||
| AK127197.1 | 2 | 1654 | ||
| AK128861.1 | ||||
| AK293057.1 | ||||
| BC020790.1 | AAH20790.1 | |||
| BQ773824.1 | ||||
| CR456941.1 | CAG33222.1 | |||
| U49245.1 | AAA91473.1 | |||
| X98001.1 | CAA66638.1 | |||
| Y08201.1 | CAA69383.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519329 | Chr.1:75364753 - 75892906 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map