Assay Details
Target Gene Details
Entrez Gene ID: | 55240 |
Gene Name: | STEAP3 metalloreductase |
Gene Aliases: |
AHMIO2, STMP3, TSAP6, dudlin-2, dudulin-2, pHyde |
Location: |
Chr.2:119222476-119265652 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STEAP3 | NM_001008410.1 | NP_001008410.1 | ||
| NM_018234.2 | NP_060704.2 | |||
| NM_138637.2 | NP_619543.2 | |||
| NM_182915.2 | NP_878919.2 | |||
| XM_006712614.3 | XP_006712677.1 | |||
| XM_006712615.1 | XP_006712678.1 | |||
| XM_011511403.1 | XP_011509705.1 | |||
| AF238864.1 | AAL78206.1 | |||
| AF262322.1 | AAM08128.1 | |||
| AF423424.1 | AAQ04065.1 | |||
| AK001691.1 | BAA91839.1 | |||
| AK024163.1 | BAB14842.1 | |||
| AK291608.1 | ||||
| AL833624.1 | CAH56204.1 | |||
| AY029585.1 | AAK50538.1 | |||
| AY082673.1 | AAM45136.1 | |||
| AY214461.1 | AAO38238.1 | |||
| BC042150.1 | AAH42150.1 | |||
| BC095421.1 | ||||
| BX538047.1 | CAD97986.1 |
Target Gene Details
Entrez Gene ID: | 100874111 |
Gene Name: | STEAP3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:119244422-119249071 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STEAP3-AS1 | NR_046721.1 | |||
| AK127773.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv582785 | Chr.2:119162879 - 119250980 on Build GRCh38 | Gain |
|
| esv3893204 | Chr.2:119240639 - 119249781 on Build GRCh38 | Loss |
|
| nsv582783 | Chr.2:118821982 - 119269323 on Build GRCh38 | Gain |
|
| nsv459119 | Chr.2:118952824 - 119514207 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map