Assay Details
Target Gene Details
Entrez Gene ID: | 159963 |
Gene Name: | solute carrier family 5 member 12 |
Gene Aliases: |
SMCT2 |
Location: |
Chr.11:26667019-26723356 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC5A12 | NM_178498.3 | 6 | 1049 | NP_848593.2 |
| XM_006718155.3 | 6 | 952 | XP_006718218.1 | |
| XM_006718156.3 | 6 | 1288 | XP_006718219.1 | |
| XM_011519920.2 | 7 | 529 | XP_011518222.1 | |
| XM_011519921.2 | 6 | 1289 | XP_011518223.1 | |
| XM_017017244.1 | 7 | 975 | XP_016872733.1 | |
| AY299482.1 | 6 | 758 | ||
| BC041454.1 | 6 | 518 | AAH41454.1 | |
| BC049207.1 | 6 | 1021 | AAH49207.1 | |
| CN370367.1 | 2 | 158 |
More Information
Set Membership: |
|

Genomic Map