Assay Details
Target Gene Details
Entrez Gene ID: | 354 |
Gene Name: | kallikrein related peptidase 3 |
Gene Aliases: |
APS, KLK2A1, PSA, hK3 |
Location: |
Chr.19:50854915-50860764 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK3 | NM_001030047.1 | 5 | 962 | NP_001025218.1 |
| NM_001030048.1 | NP_001025219.1 | |||
| NM_001648.2 | NP_001639.1 | |||
| AF335477.1 | AAL73204.1 | |||
| AF335478.1 | AAL73205.1 | |||
| AJ310938.1 | CAC41632.1 | |||
| AJ459782.1 | CAD30844.1 | |||
| AJ459783.1 | CAD30845.1 | |||
| AK312933.1 | ||||
| AX772834.1 | 4 | 784 | CAE06266.1 | |
| BC005307.1 | AAH05307.1 | |||
| BC050595.2 | AAH50595.2 | |||
| BC056665.1 | AAH56665.1 | |||
| BQ932072.1 | ||||
| BT019862.1 | AAV38665.1 | |||
| DQ641251.1 | ||||
| M21895.1 | AAA59995.1 | |||
| M21896.1 | 5 | 910 | AAA59996.1 | |
| M26663.1 | AAA58802.1 | |||
| U17040.1 | AAA56764.1 | |||
| X05332.1 | CAA28947.1 | |||
| X07730.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2531 | Chr.19:50830372 - 50875341 on Build GRCh38 | Deletion |
|
| esv2763208 | Chr.19:50858501 - 50873907 on Build GRCh38 | Gain |
|
| nsv524465 | Chr.19:50856831 - 50860142 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map