Assay Details
Target Gene Details
Entrez Gene ID: | 64429 |
Gene Name: | zinc finger DHHC-type containing 6 |
Gene Aliases: |
DHHC-6, ZNF376 |
Location: |
Chr.10:112424411-112448335 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZDHHC6 | NM_001303134.1 | 11 | 2044 | NP_001290063.1 |
| NM_022494.2 | 11 | 2056 | NP_071939.1 | |
| XM_005270051.4 | 11 | 2991 | XP_005270108.1 | |
| XM_006717948.3 | 11 | 2162 | XP_006718011.1 | |
| XM_006717949.3 | 10 | 1891 | XP_006718012.1 | |
| XM_011540091.2 | XP_011538393.1 | |||
| XM_011540092.2 | 11 | 1961 | XP_011538394.1 | |
| XM_011540093.1 | 11 | 2070 | XP_011538395.1 | |
| XM_011540094.2 | 11 | 1929 | XP_011538396.1 | |
| XM_011540095.2 | 10 | 1779 | XP_011538397.1 | |
| XM_017016561.1 | 11 | 1982 | XP_016872050.1 | |
| XM_017016562.1 | 11 | 2150 | XP_016872051.1 | |
| XM_017016563.1 | 11 | 1963 | XP_016872052.1 | |
| XM_017016564.1 | XP_016872053.1 | |||
| XM_017016565.1 | XP_016872054.1 | |||
| XM_017016566.1 | XP_016872055.1 | |||
| XM_017016567.1 | 10 | 1879 | XP_016872056.1 | |
| XM_017016568.1 | 11 | 1867 | XP_016872057.1 | |
| XM_017016569.1 | 7 | 1004 | XP_016872058.1 | |
| AI695150.1 | 1 | 176 | ||
| AK025605.1 | 11 | 2011 | BAB15187.1 | |
| AK026369.1 | 6 | 916 | BAB15462.1 | |
| AK223086.1 | 11 | 2010 | BAD96806.1 | |
| BX647270.1 | 9 | 4500 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831990 | Chr.10:112349267 - 112531997 on Build GRCh38 | Loss |
|
| nsv831991 | Chr.10:112413098 - 112582139 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map