Assay Details
Target Gene Details
Entrez Gene ID: | 25956 |
Gene Name: | SEC31 homolog B, COPII coat complex component |
Gene Aliases: |
SEC31B-1, SEC31L2 |
Location: |
Chr.10:100486646-100519838 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC31B | NM_015490.3 | NP_056305.1 | ||
| AF274863.1 | AAF78243.1 | |||
| AK128343.1 | ||||
| AL080141.1 | CAB45735.1 | |||
| BC144313.1 | ||||
| BX640914.1 | CAE45955.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818783 | Chr.10:100461667 - 100509449 on Build GRCh38 | Gain |
|
| nsv831963 | Chr.10:100458231 - 100678922 on Build GRCh38 | Loss |
|
| nsv831962 | Chr.10:100406551 - 100593251 on Build GRCh38 | Gain |
|
| nsv8723 | Chr.10:100453735 - 100510190 on Build GRCh38 | Gain |
|
| esv34217 | Chr.10:100463200 - 100515885 on Build GRCh38 | Gain |
|
| esv3624362 | Chr.10:100454267 - 100509638 on Build GRCh38 | Gain |
|
| esv2759779 | Chr.10:100392093 - 100805979 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map