Assay Details
Target Gene Details
Entrez Gene ID: | 1456 |
Gene Name: | casein kinase 1 gamma 3 |
Gene Aliases: |
CKI-gamma 3, CSNK1G3L |
Location: |
Chr.5:123512099-123617045 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSNK1G3 | NM_001031812.3 | 1 | 159 | NP_001026982.1 |
| NM_001270573.1 | 1 | 159 | NP_001257502.1 | |
| NM_001270574.1 | 1 | 159 | NP_001257503.1 | |
| XM_005271891.1 | 1 | 159 | XP_005271948.1 | |
| XM_005271892.2 | 1 | 159 | XP_005271949.1 | |
| XM_005271894.2 | 1 | 159 | XP_005271951.1 | |
| XM_005271895.2 | 1 | 159 | XP_005271952.1 | |
| XM_005271896.1 | 1 | 159 | XP_005271953.1 | |
| XM_017009057.1 | 1 | 159 | XP_016864546.1 | |
| XM_017009058.1 | 1 | 142 | XP_016864547.1 | |
| XM_017009059.1 | 1 | 142 | XP_016864548.1 | |
| XM_017009060.1 | 1 | 142 | XP_016864549.1 | |
| XM_017009061.1 | 1 | 159 | XP_016864550.1 | |
| XM_017009063.1 | 1 | 159 | XP_016864552.1 | |
| XM_017009064.1 | 1 | 159 | XP_016864553.1 | |
| XM_017009065.1 | 1 | 159 | XP_016864554.1 | |
| XM_017009066.1 | 1 | 142 | XP_016864555.1 | |
| XM_017009067.1 | 1 | 159 | XP_016864556.1 | |
| XM_017009068.1 | 1 | 159 | XP_016864557.1 | |
| XM_017009069.1 | 1 | 142 | XP_016864558.1 | |
| XM_017009070.1 | 1 | 159 | XP_016864559.1 | |
| AK299739.1 | ||||
| AK316019.1 | ||||
| CN286763.1 | 1 | 75 | ||
| DA409424.1 | 1 | 159 |
More Information
Set Membership: |
|

Genomic Map