Assay Details
Target Gene Details
Entrez Gene ID: | 115677 |
Gene Name: | nitric oxide synthase trafficking |
Gene Aliases: |
DaIP2 |
Location: |
Chr.2:168786539-168865339 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOSTRIN | NM_001039724.3 | NP_001034813.2 | ||
| NM_001171631.1 | 5 | 719 | NP_001165102.1 | |
| NM_001171632.1 | NP_001165103.1 | |||
| NM_052946.3 | NP_443178.2 | |||
| XM_005246270.1 | XP_005246327.1 | |||
| XM_006712233.3 | XP_006712296.1 | |||
| XM_011510540.2 | XP_011508842.1 | |||
| XM_017003278.1 | XP_016858767.1 | |||
| AK002203.2 | BAE46614.1 | |||
| AK093444.1 | 5 | 717 | ||
| AK290254.1 | ||||
| BC014189.2 | AAH14189.1 | |||
| BC093072.1 | AAH93072.1 | |||
| BP334580.1 |
More Information
Set Membership: |
|

Genomic Map