Assay Details
Target Gene Details
Entrez Gene ID: | 6236 |
Gene Name: | RRAD, Ras related glycolysis inhibitor and calcium channel regulator |
Gene Aliases: |
RAD, RAD1, REM3 |
Location: |
Chr.16:66921679-66925536 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRAD | NM_001128850.1 | NP_001122322.1 | ||
| NM_004165.2 | NP_004156.1 | |||
| AK290613.1 | ||||
| BC011645.2 | AAH11645.1 | |||
| BC057815.1 | AAH57815.1 | |||
| BP257026.1 | ||||
| L24564.1 | AAA36540.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827706 | Chr.16:66894354 - 67009614 on Build GRCh38 | Loss |
|
| nsv572904 | Chr.16:66876182 - 66938470 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map