Assay Details
Target Gene Details
Entrez Gene ID: | 8030 |
Gene Name: | coiled-coil domain containing 6 |
Gene Aliases: |
D10S170, H4, PTC, TPC, TST1 |
Location: |
Chr.10:59788747-59906656 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC6 | NM_005436.4 | 9 | 5231 | NP_005427.2 |
| AK024913.1 | 1 | 1430 | ||
| AK055515.1 | 1 | 2042 | ||
| BC036757.2 | 9 | 5070 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv948030 | Chr.10:59788818 - 59790404 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map