Assay Details
Target Gene Details
| Entrez Gene ID: | 152330 | 
| Gene Name: | contactin 4 | 
| Gene Aliases: | AXCAM, BIG-2 | 
| Location: | Chr.3:2098771-3059080 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 28 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CNTN4 | NM_001206955.1 | 19 | 2609 | NP_001193884.1 | 
| NM_001206956.1 | 11 | 1328 | NP_001193885.1 | |
| NM_175607.2 | 20 | 2564 | NP_783200.1 | |
| NM_175613.2 | 11 | 1331 | NP_783302.1 | |
| XM_006713004.3 | 19 | 2574 | XP_006713067.1 | |
| XM_011533425.2 | 19 | 2577 | XP_011531727.1 | |
| XM_011533426.2 | 20 | 2796 | XP_011531728.1 | |
| XM_011533427.2 | 20 | 2777 | XP_011531729.1 | |
| XM_011533428.2 | 18 | 2766 | XP_011531730.1 | |
| XM_011533429.2 | 18 | 2701 | XP_011531731.1 | |
| XM_011533430.2 | 18 | 2662 | XP_011531732.1 | |
| XM_011533431.2 | 17 | 5411 | XP_011531733.1 | |
| XM_017005782.1 | 20 | 2519 | XP_016861271.1 | |
| XM_017005783.1 | 21 | 3393 | XP_016861272.1 | |
| XM_017005784.1 | 19 | 2854 | XP_016861273.1 | |
| XM_017005785.1 | 20 | 2501 | XP_016861274.1 | |
| XM_017005786.1 | 18 | 2763 | XP_016861275.1 | |
| XM_017005787.1 | 18 | 2659 | XP_016861276.1 | |
| AF464063.1 | 1 | 475 | AAN86141.1 | |
| AF549455.1 | 17 | 2607 | AAP05786.1 | |
| AK057278.1 | 11 | 1309 | ||
| AK097633.1 | 14 | 1855 | ||
| AK125460.1 | 19 | 2752 | ||
| AK314396.1 | 11 | 1309 | ||
| AY090737.1 | 19 | 2446 | AAM00025.1 | |
| BC026119.1 | 11 | 1311 | AAH26119.1 | 
Target Gene Details
| Entrez Gene ID: | 100873975 | 
| Gene Name: | CNTN4 antisense RNA 1 | 
| Gene Aliases: | - | 
| Location: | Chr.3:3039661-3061145 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 4 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CNTN4-AS1 | NR_046554.1 | 4 | 575 | |
| BM920658.1 | 4 | 575 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv589371 | Chr.3:2170634 - 3594980 on Build GRCh38 | Loss |  CRBN  CNTN4  LOC100130207  TRNT1  CNTN4-AS1  IL5RA | 
| esv2751994 | Chr.3:2200757 - 3178886 on Build GRCh38 | Gain |  CRBN  CNTN4  TRNT1  CNTN4-AS1  IL5RA | 
| esv2422274 | Chr.3:2100396 - 3314907 on Build GRCh38 | Duplication |  CRBN  CNTN4  LOC100130207  TRNT1  CNTN4-AS2  CNTN4-AS1  IL5RA | 
| nsv1011872 | Chr.3:2782874 - 3057776 on Build GRCh38 | Gain |  CNTN4  CNTN4-AS1 | 
| nsv521796 | Chr.3:3038825 - 3151219 on Build GRCh38 | Loss |  CRBN  CNTN4  TRNT1  CNTN4-AS1  IL5RA | 
| dgv4636n100 | Chr.3:2825734 - 3271845 on Build GRCh38 | Gain |  CRBN  CNTN4  LOC100130207  TRNT1  CNTN4-AS1  IL5RA | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top