Assay Details
Target Gene Details
Entrez Gene ID: | 26094 |
Gene Name: | DDB1 and CUL4 associated factor 4 |
Gene Aliases: |
WDR21, WDR21A |
Location: |
Chr.14:72926332-72962098 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF4 | NM_001163508.1 | 1 | 92 | NP_001156980.1 |
| NM_001163509.1 | 1 | 92 | NP_001156981.1 | |
| NM_015604.3 | 1 | 92 | NP_056419.2 | |
| NM_181340.2 | 1 | 92 | NP_851937.1 | |
| NM_181341.2 | 1 | 92 | NP_851938.2 | |
| XM_017021203.1 | 1 | 87 | XP_016876692.1 | |
| XM_017021207.1 | 1 | 87 | XP_016876696.1 | |
| XM_017021208.1 | 1 | 87 | XP_016876697.1 | |
| XM_017021209.1 | 1 | 87 | XP_016876698.1 | |
| XM_017021210.1 | 1 | 87 | XP_016876699.1 | |
| XM_017021214.1 | 1 | 75 | XP_016876703.1 | |
| XM_017021215.1 | 1 | 61 | XP_016876704.1 | |
| AK297571.1 | ||||
| AK315499.1 | 1 | 92 | ||
| BX161489.1 | 1 | 47 | CAD61938.1 | |
| DC314513.1 | 1 | 92 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049819 | Chr.14:72911687 - 73164900 on Build GRCh38 | Gain |
|
| nsv1048734 | Chr.14:72817765 - 73069496 on Build GRCh38 | Gain |
|
| nsv1035460 | Chr.14:72854572 - 73897173 on Build GRCh38 | Gain |
|
| esv3892560 | Chr.14:72813150 - 72958973 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map