Assay Details
Target Gene Details
Entrez Gene ID: | 51207 |
Gene Name: | dual specificity phosphatase 13 |
Gene Aliases: |
BEDP, DUSP13A, DUSP13B, MDSP, SKRP4, TMDP |
Location: |
Chr.10:75094432-75109221 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUSP13 | NM_001007271.1 | 8 | 1687 | NP_001007272.1 |
| NM_001007272.1 | 4 | 962 | NP_001007273.1 | |
| NM_001007273.1 | 6 | 1261 | NP_001007274.1 | |
| NM_001320842.1 | 7 | 1479 | NP_001307771.1 | |
| NM_001320843.1 | 5 | 1057 | NP_001307772.1 | |
| NM_016364.3 | 4 | 820 | NP_057448.3 | |
| XM_005269883.3 | 7 | 1457 | XP_005269940.1 | |
| XM_005269884.4 | 4 | 1249 | XP_005269941.3 | |
| XM_005269887.1 | 7 | 1559 | XP_005269944.1 | |
| XM_005269890.1 | 5 | 1159 | XP_005269947.1 | |
| XM_011539853.1 | 8 | 1696 | XP_011538155.1 | |
| XM_011539854.2 | 6 | 1235 | XP_011538156.1 | |
| XM_011539855.1 | 4 | 938 | XP_011538157.1 | |
| XM_011539856.2 | 6 | 1239 | XP_011538158.1 | |
| XM_017016313.1 | 4 | 912 | XP_016871802.1 | |
| AB027004.1 | 4 | 834 | BAA89412.1 | |
| AB103375.1 | 4 | 834 | BAD91014.1 | |
| AI971328.1 | 1 | 102 | ||
| AK057012.1 | 5 | 1030 | ||
| AK127886.1 | 7 | 1479 | ||
| AY040091.1 | 8 | 1651 | AAK77966.1 | |
| AY674051.1 | 8 | 1660 | AAT79356.1 | |
| BC009778.1 | 5 | 946 | AAH09778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv927n100 | Chr.10:75024700 - 75329039 on Build GRCh38 | Gain |
|
| nsv7199 | Chr.10:75077582 - 75112320 on Build GRCh38 | Insertion |
|
| nsv518639 | Chr.10:75083167 - 75108141 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map