Assay Details
Target Gene Details
Entrez Gene ID: | 374407 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B13 |
Gene Aliases: |
RSPH16A, TSARG5, TSARG6 |
Location: |
Chr.11:73950319-73970287 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB13 | NM_153614.3 | 6 | 1410 | NP_705842.2 |
| XM_005273984.3 | 5 | 775 | XP_005274041.1 | |
| XM_011545004.2 | 8 | 1661 | XP_011543306.1 | |
| XM_011545005.2 | 7 | 1501 | XP_011543307.1 | |
| XM_011545007.2 | 6 | 1186 | XP_011543309.1 | |
| XM_011545009.2 | 8 | 1335 | XP_011543311.1 | |
| XM_011545015.2 | 3 | 528 | XP_011543317.1 | |
| XM_017017675.1 | 7 | 931 | XP_016873164.1 | |
| XM_017017676.1 | 8 | 1511 | XP_016873165.1 | |
| XM_017017677.1 | 7 | 1380 | XP_016873166.1 | |
| XM_017017678.1 | 6 | 1191 | XP_016873167.1 | |
| AF516185.1 | 3 | 1382 | AAP47195.1 | |
| AK058095.1 | 3 | 1382 | ||
| AK128589.1 | 8 | 3455 | ||
| AY138810.2 | 6 | 1410 | AAN15929.1 | |
| AY325766.1 | 6 | 703 |
More Information
Set Membership: |
|

Genomic Map