Assay Details
Target Gene Details
Entrez Gene ID: | 9760 |
Gene Name: | thymocyte selection associated high mobility group box |
Gene Aliases: |
TOX1 |
Location: |
Chr.8:58805418-59119293 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOX | NM_014729.2 | 9 | 3371 | NP_055544.1 |
| XM_017014085.1 | 8 | 3390 | XP_016869574.1 | |
| AB018351.1 | 9 | 3373 | ||
| CA439630.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6213 | Chr.8:58790507 - 58824628 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map