Assay Details
Target Gene Details
Entrez Gene ID: | 146562 |
Gene Name: | chromosome 16 open reading frame 71 |
Gene Aliases: |
- |
Location: |
Chr.16:4734288-4749396 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C16orf71 | NM_139170.2 | 10 | 2230 | NP_631909.2 |
| XM_005255144.3 | 9 | 1959 | XP_005255201.1 | |
| XM_017022975.1 | 10 | 2086 | XP_016878464.1 | |
| XM_017022976.1 | 10 | 1848 | XP_016878465.1 | |
| AF447587.1 | 10 | 2204 | AAM22870.1 | |
| AK125251.1 | 7 | 1259 | ||
| AK301794.1 | ||||
| BC035024.1 | 10 | 2180 | AAH35024.1 | |
| BX648126.1 | 8 | 3229 |
Target Gene Details
Entrez Gene ID: | 26048 |
Gene Name: | zinc finger protein 500 |
Gene Aliases: |
ZKSCAN18, ZSCAN50 |
Location: |
Chr.16:4745961-4767219 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF500 | NM_021646.3 | 6 | 5199 | NP_067678.1 |
| XM_011522453.2 | XP_011520755.1 | |||
| AB011129.1 | 5 | 4939 | ||
| BM678198.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| nsv571302 | Chr.16:4708464 - 4762815 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map