Assay Details
Target Gene Details
Entrez Gene ID: | 1687 |
Gene Name: | DFNA5, deafness associated tumor suppressor |
Gene Aliases: |
ICERE-1 |
Location: |
Chr.7:24698355-24758020 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DFNA5 | NM_001127453.1 | 10 | 1972 | NP_001120925.1 |
| NM_001127454.1 | 9 | 1987 | NP_001120926.1 | |
| NM_004403.2 | 10 | 2217 | NP_004394.1 | |
| XM_017011802.1 | 9 | 1743 | XP_016867291.1 | |
| AF007790.2 | 9 | 1872 | AAC39635.2 | |
| AF073308.1 | 10 | 1941 | AAC69324.1 | |
| AF075171.1 | 2 | 1626 | AAC69326.1 | |
| AF131765.1 | 2 | 1628 | AAD20039.1 | |
| AJ270686.1 | 1 | 242 | ||
| AK094714.1 | 12 | 2545 | ||
| BC099911.1 | 10 | 1933 | ||
| BC125065.1 | ||||
| BC125066.1 | ||||
| BX647389.1 | 2 | 3904 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3612588 | Chr.7:24698120 - 24699524 on Build GRCh38 | Loss |
|
| nsv520276 | Chr.7:24649588 - 25027423 on Build GRCh38 | Gain |
|
| nsv527599 | Chr.7:24591626 - 24708291 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map