Assay Details
Target Gene Details
Entrez Gene ID: | 9715 |
Gene Name: | family with sequence similarity 131 member B |
Gene Aliases: |
- |
Location: |
Chr.7:143353400-143382304 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM131B | NM_001031690.2 | 2 | 230 | NP_001026860.2 |
| NM_001278297.1 | 2 | 230 | NP_001265226.1 | |
| NM_014690.4 | 2 | 123 | NP_055505.3 | |
| XM_005250073.3 | 2 | 444 | XP_005250130.1 | |
| XM_006716186.2 | 2 | 123 | XP_006716249.1 | |
| AK124557.1 | 2 | 178 | ||
| AK291470.1 | 2 | 95 | ||
| AL832579.2 | 1 | 1714 | CAD89938.1 | |
| BC045611.1 | 2 | 235 | AAH45611.2 | |
| BC050543.1 | 2 | 98 | ||
| DA226407.1 | 2 | 123 | ||
| DA334424.1 | 2 | 115 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517520 | Chr.7:143352880 - 143400007 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map