Assay Details
Target Gene Details
Entrez Gene ID: | 390321 |
Gene Name: | olfactory receptor family 6 subfamily C member 1 |
Gene Aliases: |
OST267 |
Location: |
Chr.12:55320600-55321538 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6C1 | NM_001005182.1 | 1 | 223 | NP_001005182.1 |
| BC136957.1 | 1 | 262 | ||
| BC136965.1 | 1 | 261 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv82 | Chr.12:55315883 - 55331656 on Build GRCh38 | Insertion |
|
| nsv559002 | Chr.12:55319822 - 55322580 on Build GRCh38 | Loss |
|
| nsv1050720 | Chr.12:55313191 - 55326825 on Build GRCh38 | Loss |
|
| esv3629571 | Chr.12:55310583 - 55331520 on Build GRCh38 | Loss |
|
| nsv559001 | Chr.12:55113681 - 55663924 on Build GRCh38 | Loss |
|
| esv2761740 | Chr.12:55313203 - 55326837 on Build GRCh38 | Loss |
|
| nsv718 | Chr.12:55312572 - 55352417 on Build GRCh38 | Insertion |
|
| nsv826381 | Chr.12:55276757 - 55331791 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75802465] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map