Assay Details
Target Gene Details
Entrez Gene ID: | 147650 |
Gene Name: | sperm acrosome associated 6 |
Gene Aliases: |
LET7EH, LINC00085, NCRNA00085, SPACA6P |
Location: |
Chr.19:51688211-51712071 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPACA6 | NM_001316972.1 | NP_001303901.1 | ||
| NM_001316994.1 | NP_001303923.1 | |||
| NR_024330.1 | ||||
| XM_017026299.1 | XP_016881788.1 | |||
| XM_017026300.1 | XP_016881789.1 | |||
| XM_017026301.1 | XP_016881790.1 | |||
| AY358799.1 | AAQ89159.1 | |||
| KF751883.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9742 | Chr.19:51684285 - 51718489 on Build GRCh38 | Gain+Loss |
|
| nsv953608 | Chr.19:51689148 - 51704247 on Build GRCh38 | Deletion |
|
| esv2758767 | Chr.19:51488642 - 52101372 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76526663] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map