Assay Details
Target Gene Details
Entrez Gene ID: | 283284 |
Gene Name: | immunoglobulin superfamily member 22 |
Gene Aliases: |
IGFN2 |
Location: |
Chr.11:18704305-18726939 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF22 | NM_173588.3 | 23 | 4242 | NP_775859.3 |
| XM_011520030.2 | 21 | 7146 | XP_011518332.1 | |
| XM_011520031.2 | 20 | 6843 | XP_011518333.1 | |
| XM_017017607.1 | 22 | 7037 | XP_016873096.1 | |
| XM_017017608.1 | 22 | 5060 | XP_016873097.1 | |
| AA255875.1 | 1 | 119 | ||
| AI952274.1 | 1 | 122 | ||
| AK095113.1 | 21 | 3850 | BAC04488.1 |
Target Gene Details
Entrez Gene ID: | 144110 |
Gene Name: | transmembrane protein 86A |
Gene Aliases: |
- |
Location: |
Chr.11:18698735-18704785 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM86A | NM_153347.2 | 3 | 3263 | NP_699178.1 |
| BC035692.1 | 3 | 3194 | AAH35692.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553633 | Chr.11:18290154 - 19036904 on Build GRCh38 | Gain |
|
| nsv525192 | Chr.11:18701244 - 18771813 on Build GRCh38 | Loss |
|

Genomic Map