Assay Details
Target Gene Details
Entrez Gene ID: | 1583 |
Gene Name: | cytochrome P450 family 11 subfamily A member 1 |
Gene Aliases: |
CYP11A, CYPXIA1, P450SCC |
Location: |
Chr.15:74337762-74367740 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP11A1 | NM_000781.2 | 9 | 1673 | NP_000772.2 |
| NM_001099773.1 | 9 | 1747 | NP_001093243.1 | |
| AI149078.1 | 1 | 264 | ||
| AK056794.1 | 9 | 1742 | ||
| AK225715.1 | 8 | 1261 | ||
| AK292300.1 | 9 | 1579 | ||
| AK292301.1 | 8 | 1537 | ||
| AK293211.1 | 10 | 1539 | ||
| AK300178.1 | 9 | 1489 | ||
| BC032329.1 | 9 | 1562 | AAH32329.1 | |
| KU178011.1 | ||||
| KU178012.1 | ||||
| KU178013.1 | ||||
| KU178014.1 | ||||
| M14565.1 | 9 | 1562 | AAA52162.1 | |
| M28253.1 | 5 | 672 | AAA36404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636903 | Chr.15:74336145 - 74342188 on Build GRCh38 | Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map