Assay Details
Target Gene Details
Entrez Gene ID: | 29925 |
Gene Name: | GDP-mannose pyrophosphorylase B |
Gene Aliases: |
MDDGA14, MDDGB14, MDDGC14 |
Location: |
Chr.3:49721476-49723974 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GMPPB | NM_013334.3 | NP_037466.2 | ||
| NM_021971.2 | NP_068806.1 | |||
| AF135421.1 | AAD38516.1 | |||
| AK024319.1 | BAB14882.1 | |||
| AK291700.1 | ||||
| BC001141.1 | AAH01141.1 | |||
| BC008033.1 | AAH08033.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508217 | Chr.3:49578013 - 49732463 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map