Assay Details
Target Gene Details
Entrez Gene ID: | 54475 |
Gene Name: | notchless homolog 1 |
Gene Aliases: |
Nle |
Location: |
Chr.17:35128755-35142315 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLE1 | NM_001014445.2 | 12 | 2569 | NP_001014445.1 |
| NM_018096.4 | 13 | 2498 | NP_060566.2 | |
| XM_017024777.1 | 13 | 2516 | XP_016880266.1 | |
| AB209111.1 | 12 | 2569 | BAD92348.1 | |
| AJ005257.1 | 4 | 1263 | CAA06444.1 | |
| AW451963.1 | 1 | 106 | ||
| BC012075.1 | 13 | 2471 | AAH12075.1 | |
| HY320157.1 | 1 | 147 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2036 | Chr.17:35124777 - 35143140 on Build GRCh38 | Insertion |
|
| esv3892998 | Chr.17:35106468 - 35295755 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map