Assay Details
Target Gene Details
Entrez Gene ID: | 5710 |
Gene Name: | proteasome 26S subunit, non-ATPase 4 |
Gene Aliases: |
AF, AF-1, ASF, MCB1, Rpn10, S5A, pUB-R5 |
Location: |
Chr.1:151254700-151267479 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMD4 | NM_002810.2 | NP_002801.1 | ||
| XM_005245354.2 | 1 | 33 | XP_005245411.1 | |
| XM_006711464.3 | XP_006711527.1 | |||
| XM_017001856.1 | XP_016857345.1 | |||
| AB033605.1 | BAA97581.1 | |||
| BC002365.2 | AAH02365.1 | |||
| BC072008.1 | AAH72008.1 | |||
| U24704.1 | AAB54057.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| esv3587522 | Chr.1:151237373 - 151267334 on Build GRCh38 | Gain |
|
| dgv2n68 | Chr.1:151136983 - 151309302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map