Assay Details
Target Gene Details
Entrez Gene ID: | 79639 |
Gene Name: | transmembrane protein 53 |
Gene Aliases: |
NET4 |
Location: |
Chr.1:44653247-44674608 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM53 | NM_001300746.1 | 2 | 941 | NP_001287675.1 |
| NM_001300747.1 | 3 | 973 | NP_001287676.1 | |
| NM_001300748.1 | 3 | 970 | NP_001287677.1 | |
| NM_024587.3 | 3 | 1063 | NP_078863.2 | |
| XM_011542138.2 | 3 | 1192 | XP_011540440.1 | |
| AK026006.1 | 3 | 870 | BAB15317.1 | |
| AK225405.1 | 3 | 870 | ||
| AK295356.1 | 2 | 663 | ||
| AK296548.1 | 2 | 779 | ||
| AK310016.1 | 3 | 1007 | ||
| AL832539.1 | 2 | 775 | CAI46125.1 | |
| BC064520.1 | 3 | 882 | AAH64520.1 | |
| BM806701.1 | ||||
| CR457348.1 | CAG33629.1 |

Genomic Map