Assay Details
Target Gene Details
Entrez Gene ID: | 1111 |
Gene Name: | checkpoint kinase 1 |
Gene Aliases: |
CHK1 |
Location: |
Chr.11:125624910-125676259 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHEK1 | NM_001114121.2 | NP_001107593.1 | ||
| NM_001114122.2 | NP_001107594.1 | |||
| NM_001244846.1 | NP_001231775.1 | |||
| NM_001274.5 | NP_001265.2 | |||
| NR_045204.1 | ||||
| NR_045205.1 | ||||
| XM_011542560.2 | XP_011540862.1 | |||
| XM_011542561.2 | XP_011540863.1 | |||
| XM_011542562.2 | XP_011540864.1 | |||
| XM_011542563.2 | XP_011540865.1 | |||
| XM_017017146.1 | XP_016872635.1 | |||
| AB451222.1 | ||||
| AB451345.1 | ||||
| AF016582.1 | AAC51736.1 | |||
| AF032874.1 | AAB88852.1 | |||
| AK292549.1 | ||||
| AK293143.1 | ||||
| AK299783.1 | ||||
| AK300082.1 | ||||
| BC004202.2 | AAH04202.1 | |||
| BC017575.1 | AAH17575.1 | |||
| BG468734.1 | ||||
| BP207812.1 | ||||
| DA567616.1 | ||||
| JF289264.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map