Assay Details
Target Gene Details
Entrez Gene ID: | 81341 |
Gene Name: | olfactory receptor family 10 subfamily W member 1 |
Gene Aliases: |
OR10W1P, OR10W1Q, OR11-236, UNQ6469 |
Location: |
Chr.11:58266792-58268260 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10W1 | NM_207374.3 | 1 | 1231 | NP_997257.2 |
| AY358251.1 | 1 | 1231 | AAQ88618.1 | |
| BC132848.1 | 1 | 1130 | ||
| BC132850.1 | 1 | 1130 |
More Information
Set Membership: |
|

Genomic Map