Assay Details
Target Gene Details
Entrez Gene ID: | 7701 |
Gene Name: | zinc finger protein 142 |
Gene Aliases: |
pHZ-49 |
Location: |
Chr.2:218637592-218659632 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF142 | NM_001105537.2 | 8 | 1974 | NP_001099007.1 |
| NR_073599.1 | 9 | 2283 | ||
| NR_073600.1 | 9 | 2116 | ||
| XM_006712730.3 | 9 | 2649 | XP_006712793.1 | |
| XM_011511784.2 | 9 | 2744 | XP_011510086.1 | |
| XM_011511785.2 | 9 | 2669 | XP_011510087.1 | |
| XM_011511786.2 | 9 | 2574 | XP_011510088.1 | |
| XM_011511787.2 | 8 | 2569 | XP_011510089.1 | |
| XM_011511788.2 | 8 | 2474 | XP_011510090.1 | |
| XM_011511789.2 | 8 | 2142 | XP_011510091.1 | |
| XM_017004871.1 | 8 | 2725 | XP_016860360.1 | |
| XM_017004872.1 | 7 | 2125 | XP_016860361.1 | |
| AK095027.1 | ||||
| D87073.1 | 8 | 1972 | ||
| U09849.1 | 2 | 159 | AAC50265.1 |
More Information
Set Membership: |
|

Genomic Map