Assay Details
Target Gene Details
Entrez Gene ID: | 10663 |
Gene Name: | C-X-C motif chemokine receptor 6 |
Gene Aliases: |
BONZO, CD186, STRL33, TYMSTR |
Location: |
Chr.3:45940688-45948354 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCR6 | NM_006564.1 | 2 | 1262 | NP_006555.1 |
| XM_005264809.2 | 2 | 1336 | XP_005264866.1 | |
| XM_011533290.2 | 2 | 1861 | XP_011531592.1 | |
| XM_011533291.2 | 2 | 1812 | XP_011531593.1 | |
| BC033584.1 | 1 | 1213 | AAH33584.1 | |
| JC209412.1 | 2 | 1262 | ||
| U73529.1 | 1 | 1211 | AAB61456.1 | |
| U73531.1 | 1 | 2644 | AAB61457.1 | |
| Y13248.1 | 2 | 1262 | CAA73698.1 |
Target Gene Details
Entrez Gene ID: | 79443 |
Gene Name: | FYVE and coiled-coil domain containing 1 |
Gene Aliases: |
CATC2, CTRCT18, RUFY3, ZFYVE7 |
Location: |
Chr.3:45917899-45995824 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FYCO1 | NM_024513.3 | NP_078789.2 | ||
| XM_006713333.3 | XP_006713396.1 | |||
| XM_006713334.3 | XP_006713397.1 | |||
| XM_011534111.2 | XP_011532413.1 | |||
| AJ292348.2 | CAC33883.1 | |||
| AK023397.1 | BAB14559.1 | |||
| AK074165.1 | BAB84991.1 | |||
| AL832358.1 | CAD91151.1 | |||
| AL833308.2 | CAD89924.1 | |||
| BC007218.1 | AAH07218.1 | |||
| BC101468.1 | AAI01469.1 | |||
| BC101470.1 | AAI01471.1 | |||
| BC143368.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834678 | Chr.3:45762802 - 45968764 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map