Assay Details
Target Gene Details
Entrez Gene ID: | 9334 |
Gene Name: | beta-1,4-galactosyltransferase 5 |
Gene Aliases: |
B4Gal-T5, BETA4-GALT-IV, beta4Gal-T5, beta4GalT-V, gt-V |
Location: |
Chr.20:49632945-49713884 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALT5 | NM_004776.3 | 9 | 1585 | NP_004767.1 |
| AB004550.1 | 9 | 1503 | BAA25006.1 | |
| AF038663.1 | 9 | 1401 | AAC39736.1 | |
| BC104987.1 | 9 | 1424 | AAI04988.1 | |
| BC112265.1 | 9 | 1424 | AAI12266.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3646018 | Chr.20:49621782 - 49646113 on Build GRCh38 | Gain |
|
| nsv1065852 | Chr.20:49502654 - 49643666 on Build GRCh38 | Loss |
|
| esv3646017 | Chr.20:49610386 - 49857106 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map