Assay Details
Target Gene Details
Entrez Gene ID: | 5255 |
Gene Name: | phosphorylase kinase regulatory subunit alpha 1 |
Gene Aliases: |
PHKA |
Location: |
Chr.X:72578814-72714181 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHKA1 | NM_001122670.1 | 31 | 3938 | NP_001116142.1 |
| NM_001172436.1 | 30 | 3761 | NP_001165907.1 | |
| NM_002637.3 | 32 | 3977 | NP_002628.2 | |
| XM_006724661.2 | 31 | 3642 | XP_006724724.1 | |
| AK123357.1 | 4 | 2547 | ||
| BC104944.1 | 31 | 3684 | AAI04945.1 | |
| BC143499.1 | 31 | 3684 | ||
| BC143501.1 | 30 | 3507 | ||
| DA171597.1 | 3 | 445 | ||
| X73874.1 | 32 | 3837 | CAA52083.1 |
More Information
Set Membership: |
|

Genomic Map