Assay Details
Target Gene Details
Entrez Gene ID: | 10726 |
Gene Name: | nuclear distribution C, dynein complex regulator |
Gene Aliases: |
HNUDC, MNUDC, NPD011 |
Location: |
Chr.1:26911128-26946871 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDC | NM_006600.3 | NP_006591.1 | ||
| XM_011540529.1 | XP_011538831.1 | |||
| XM_011540530.1 | XP_011538832.1 | |||
| XM_017000094.1 | XP_016855583.1 | |||
| XM_017000095.1 | XP_016855584.1 | |||
| XM_017000096.1 | XP_016855585.1 | |||
| AB019408.1 | BAA76628.1 | |||
| AF086922.1 | AAP97152.1 | |||
| AF100760.1 | AAD43024.1 | |||
| AF125465.1 | AAD39921.1 | |||
| AF130736.1 | AAD30517.1 | |||
| AF241788.1 | AAG44476.1 | |||
| AL136725.1 | CAB66659.1 | |||
| BC002399.2 | AAH02399.1 | |||
| BC003132.1 | AAH03132.1 | |||
| BC006147.1 | AAH06147.1 | |||
| BC007280.1 | AAH07280.1 | |||
| BC015153.1 | AAH15153.1 | |||
| BC021139.1 | AAH21139.1 | |||
| BP348144.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508689 | Chr.1:26912462 - 26959506 on Build GRCh38 | Deletion |
|
| nsv1000012 | Chr.1:26714969 - 27141528 on Build GRCh38 | Gain |
|
| nsv834524 | Chr.1:26830184 - 26994502 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map