Assay Details
Target Gene Details
Entrez Gene ID: | 22936 |
Gene Name: | elongation factor for RNA polymerase II 2 |
Gene Aliases: |
- |
Location: |
Chr.5:95885098-95962288 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELL2 | NM_012081.5 | NP_036213.2 | ||
| XM_006714575.2 | XP_006714638.1 | |||
| XM_017009239.1 | XP_016864728.1 | |||
| XM_017009240.1 | XP_016864729.1 | |||
| XM_017009241.1 | XP_016864730.1 | |||
| XM_017009242.1 | XP_016864731.1 | |||
| XM_017009243.1 | XP_016864732.1 | |||
| AB209343.1 | BAD92580.1 | |||
| AK296870.1 | ||||
| AK297956.1 | ||||
| AK300310.1 | ||||
| BC028412.2 | AAH28412.1 | |||
| BX538289.1 | CAD98084.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015180 | Chr.5:95798653 - 95950742 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map